missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylamine 99.0+%, TCI America™

Catalog No. C088725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C088725ML 25 mL
C0887500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C088725ML Supplier TCI America Supplier No. C088725ML

Chemical Identifiers

CAS 1003-03-8
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00001380
InChI Key NISGSNTVMOOSJQ-UHFFFAOYSA-N
Synonym cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine
PubChem CID 2906
IUPAC Name cyclopentanamine
SMILES NC1CCCC1

Specifications

Melting Point -86°C
Color Colorless
Boiling Point 107°C
Quantity 25 mL
UN Number 2733
Formula Weight 85.15
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclopentylamine
RTECSNumber GY8452000
TSCA Yes