missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylamine, 99%

Catalog No. AAL0196614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0196614 25 g
AAL0196622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0196614 Supplier Thermo Scientific Chemicals Supplier No. L0196614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1003-03-8 | C5H11N | 85.15 g/mol

Cyclopentylamine acts as an important raw material and intermediate used in organic synthesis and pharmaceuticals. Further, it serves as chemokine receptor 2 antagonists. It acts as a ligand in coordination chemistry and forms cis-dichlorobis(cyclopentylamine)platinum(II) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentylamine acts as an important raw material and intermediate used in organic synthesis and pharmaceuticals. Further, it serves as chemokine receptor 2 antagonists. It acts as a ligand in coordination chemistry and forms cis-dichlorobis(cyclopentylamine)platinum(II) complex.

Solubility
Miscible with water, acetone and benzene.

Notes
Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides and carbon dioxide.

Chemical Identifiers

CAS 1003-03-8
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00001380
InChI Key NISGSNTVMOOSJQ-UHFFFAOYSA-N
Synonym cyclopentylamine, aminocyclopentane, amino cyclopentane, unii-4259vry3gn, 1cb, cylcopentylamine, cyclopentyl amine, cyclopentyl-amine, n-cyclopentylamine, cyclo-pentyl amine
PubChem CID 2906
IUPAC Name cyclopentanamine
SMILES NC1CCCC1

Specifications

Melting Point -85°C
Density 0.862
pH 12.5
Boiling Point 106°C to 108°C
Flash Point 12°C (53°F)
Odor Amine-like
Refractive Index 1.4505
Quantity 25 g
UN Number UN3286
Beilstein 635706
Solubility Information Miscible with water,acetone and benzene.
Formula Weight 85.15
Percent Purity 99%
Chemical Name or Material Cyclopentylamine
Show More Show Less
Hazard Category H225-H301-H314-H317-H332-H335
Hazard Statement GHS H Statement
H225-H300-H331-H314-H318-H317
Highly flammable liquid and vapor.
Fatal if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
EINECSNumber 213-697-3
RTECSNumber GY8452000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only