missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylacetyl chloride, 97%

Catalog No. AAL1456214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1456214 25 g
AAL1456206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1456214 Supplier Thermo Scientific Chemicals Supplier No. L1456214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1122-99-2 | C7H11ClO | 146.61 g/mol

Cyclopentylacetyl chloride is used as a pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentylacetyl chloride is used as a pharmaceutical intermediate and in chemical research.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with bases and oxidizing agents. Store under inert gas.

Chemical Identifiers

CAS 1122-99-2
Molecular Formula C7H11ClO
Molecular Weight (g/mol) 146.61
MDL Number MFCD00236031
InChI Key NILLIUYSJFTTRH-UHFFFAOYSA-N
Synonym cyclopentylacetyl chloride, cyclopentaneacetyl chloride, cyclopentylacetylchloride, cylopentylacetyl chloride, acmc-1brgh, cyclopentaneacetylchloride, cyclopentylacetic chloride, cyclopentanacetyl chloride, cyclopentyl acetyl chloride, cyclopentyl-acetyl chloride
PubChem CID 3485182
IUPAC Name 2-cyclopentylacetyl chloride
SMILES ClC(=O)CC1CCCC1

Specifications

Density 1.087
Boiling Point 186°C
Flash Point 71°C (160°F)
Odor Pungent
Refractive Index 1.4625
Quantity 25 g
UN Number UN3265
Beilstein 1906113
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 146.62
Percent Purity 97%
Chemical Name or Material Cyclopentylacetyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only