missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylacetic Acid 98.0+%, TCI America™

Catalog No. p-7124669
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
C08945G 5 g
C089425G 25 g
2 options

Catalog No. C08945G

Supplier: TCI America C08945G

Only null left

Chemical Identifiers

CAS 1123-00-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001387
InChI Key YVHAIVPPUIZFBA-UHFFFAOYSA-N
Synonym cyclopentylacetic acid, cyclopentaneacetic acid, cycylopentylacetic acid, carboxymethyl cyclopentane, cyclopentyl acetic acid, cyclopentyacetic acid, cyclopentanacetic acid, cyclopentyl-acetic acid, 2-cyclopentylaceticacid, cyclopentane acetic acid
PubChem CID 71606
IUPAC Name 2-cyclopentylacetic acid
SMILES OC(=O)CC1CCCC1

Specifications

Color Yellow
Boiling Point 231°C
Quantity 5 g
Formula Weight 128.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclopentylacetic Acid