missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl isothiocyanate, 97%, Thermo Scientific™

Catalog No. TL00485DA
Change view
Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
TL00485DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00485DA Supplier Thermo Scientific Chemicals Supplier No. TL00485DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33522-03-1
Molecular Formula C6H9NS
Molecular Weight (g/mol) 127.205
MDL Number MFCD00040877
InChI Key PJOODZCPFADLCI-UHFFFAOYSA-N
Synonym cyclopentyl isothiocyanate, cyclopentane, isothiocyanato, isothiocyanato-cyclopentane, cyclopentanisothiocyanate, cyclopentylisothiocyanate, acmc-20albz
PubChem CID 141792
IUPAC Name isothiocyanatocyclopentane
SMILES C1CCC(C1)N=C=S

Specifications

CAS Min % 97.0
Density 1.03g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula C5H9NCS
Quantity 1g
Formula Weight 127.21
Percent Purity 97%
Chemical Name or Material Cyclopentyl isothiocyanate
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.