missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl Isocyanate 98.0+%, TCI America™

Catalog No. C25435G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25435G 5 g
C25431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25435G Supplier TCI America Supplier No. C25435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4747-71-1
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.144
MDL Number MFCD02093695
InChI Key CZALJDQHONFVFU-UHFFFAOYSA-N
Synonym cyclopentyl isocyanate, cyclopentylisocyanate, cyclopentane, isocyanato, isocyanic acid cyclopentyl ester, isocyanato cyclopentane, isocyanato-cyclopentane, acmc-1aspa, ksc497s6f
PubChem CID 643500
IUPAC Name isocyanatocyclopentane
SMILES C1CCC(C1)N=C=O

Specifications

Color Yellow
Boiling Point 146°C
Quantity 5 g
UN Number 3080
Formula Weight 111.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentyl Isocyanate
TSCA No
Recommended Storage Refrigerator