missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentene 95.0+%, TCI America™
Supplier: TCI America C1617500ML
Specifications
| Cyclopentene | |
| -135°C | |
| 46°C | |
| MFCD00001394 | |
| 2246 | |
| LPIQUOYDBNQMRZ-UHFFFAOYSA-N | |
| cyclopentene | |
| 8882 | |
| 68.12 | |
| Liquid |
| 142-29-0 | |
| Colorless | |
| C5H8 | |
| 500 mL | |
| 1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5 | |
| C1CC=CC1 | |
| 68.119 | |
| CHEBI:49155 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 142-29-0 | |
| 68.119 | |
| LPIQUOYDBNQMRZ-UHFFFAOYSA-N | |
| 8882 | |
| cyclopentene |
| C5H8 | |
| MFCD00001394 | |
| 1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5 | |
| CHEBI:49155 | |
| C1CC=CC1 |
Safety and Handling
EINECSNumber : (3)-3439
RTECSNumber : GY5950000
TSCA : Yes
Recommended Storage : Refrigerator