missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentene 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | -135°C |
| Color | Colorless |
| Boiling Point | 46°C |
| Quantity | 100 mL |
| UN Number | 2246 |
| Formula Weight | 68.12 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Cyclopentene |
Safety and Handling
| EINECSNumber | (3)-3439 |
| RTECSNumber | GY5950000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |