missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentene 98.0+%, TCI America™

Catalog No. p-7124479
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Catalog No. Quantity
C051125ML 25 mL
C0511100ML 100 mL
C0511500ML 500 mL
3 options

Catalog No. C051125ML

Supplier: TCI America C051125ML

Only null left

Chemical Identifiers

CAS 142-29-0
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00001394
InChI Key LPIQUOYDBNQMRZ-UHFFFAOYSA-N
Synonym 1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5
PubChem CID 8882
ChEBI CHEBI:49155
IUPAC Name cyclopentene
SMILES C1CC=CC1

Specifications

Melting Point -135°C
Color Colorless
Boiling Point 46°C
Quantity 25 mL
UN Number 2246
Formula Weight 68.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentene
EINECSNumber (3)-3439
RTECSNumber GY5950000
TSCA Yes
Recommended Storage Refrigerator