missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentene 98.0+%, TCI America™

Catalog No. C0511500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0511500ML 500 mL
C051125ML 25 mL
C0511100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C0511500ML Supplier TCI America Supplier No. C0511500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-29-0
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
MDL Number MFCD00001394
InChI Key LPIQUOYDBNQMRZ-UHFFFAOYSA-N
Synonym 1-cyclopentene, unii-onm2ckv81z, onm2ckv81z, 1-cyclopentene #, cyclopentene, cyclopent-2-en-1-yl, cyclopent-3-en-1-yl, dsstox_cid_9171, wln: l5utj, acmc-209co5
PubChem CID 8882
ChEBI CHEBI:49155
IUPAC Name cyclopentene
SMILES C1CC=CC1

Specifications

Melting Point -135°C
Color Colorless
Boiling Point 46°C
Quantity 500 mL
UN Number 2246
Formula Weight 68.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentene
EINECSNumber (3)-3439
RTECSNumber GY5950000
TSCA Yes
Recommended Storage Refrigerator