missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentene oxide, 97%

Catalog No. p-7025242
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2062914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2062914 Supplier Thermo Scientific Chemicals Supplier No. B2062914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 285-67-6 | C5H8O | 84.12 g/mol

Cyclopentene oxide is used to prepare beta-amino alcohols by reacting with aromatic amines using bismuth trichloride as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentene oxide is used to prepare beta-amino alcohols by reacting with aromatic amines using bismuth trichloride as a catalyst.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with acids, bases and oxidizing agents.

Chemical Identifiers

CAS 285-67-6
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00005161
InChI Key GJEZBVHHZQAEDB-UHFFFAOYNA-N
Synonym cyclopentene oxide, 6-oxabicyclo 3.1.0 hexane, 1,2-epoxycyclopentane, cyclopentane oxide, cyclopentene epoxide, epoxycyclopentane, cis-1,2-epoxycyclopentane, cyclopentane, 1,2-epoxy, 6-oxa-bicyclo 3.1.0 hexane
PubChem CID 9244
IUPAC Name 6-oxabicyclo[3.1.0]hexane
SMILES C1CC2OC2C1

Specifications

Density 0.965
Boiling Point 101°C to 102°C
Flash Point 10°C (50°F)
Refractive Index 1.434
Quantity 25 g
UN Number UN1993
Beilstein 102495
Solubility Information Immiscible with water.
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material Cyclopentene oxide
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H341-H315-H319-H335
Highly flammable liquid and vapour.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 206-005-6
RTECSNumber RN8935000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only