missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanone 99.0+%, TCI America™

Catalog No. p-7124478
Change view
Click to view available options
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
C051025ML 25 mL
C0510100ML 100 mL
C0510500ML 500 mL
3 options

Catalog No. C051025ML

Supplier: TCI America C051025ML

Only null left

Chemical Identifiers

CAS 120-92-3
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001409
InChI Key BGTOWKSIORTVQH-UHFFFAOYSA-N
Synonym adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy
PubChem CID 8452
ChEBI CHEBI:16486
IUPAC Name cyclopentanone
SMILES O=C1CCCC1

Specifications

Melting Point -51°C
Color Colorless
Boiling Point 130°C
Quantity 25 mL
UN Number 2245
Formula Weight 84.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentanone
EINECSNumber (9)-2108
RTECSNumber GY4725000
TSCA Yes