missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanone 99.0+%, TCI America™

Catalog No. C051025ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C051025ML 25 mL
C0510100ML 100 mL
C0510500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C051025ML Supplier TCI America Supplier No. C051025ML

Chemical Identifiers

CAS 120-92-3
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001409
InChI Key BGTOWKSIORTVQH-UHFFFAOYSA-N
Synonym adipic ketone, ketocyclopentane, ketopentamethylene, adipinketon, dumasin, cyclopentanon, oxocyclopentane, cyclopentan-1-one, unii-220w81tn3s, cyclopentyloxy
PubChem CID 8452
ChEBI CHEBI:16486
IUPAC Name cyclopentanone
SMILES O=C1CCCC1

Specifications

Melting Point -51°C
Color Colorless
Boiling Point 130°C
Quantity 25 mL
UN Number 2245
Formula Weight 84.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentanone
EINECSNumber (9)-2108
RTECSNumber GY4725000
TSCA Yes