missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxaldehyde (stabilized with HQ) 90.0+%, TCI America™

Supplier:  TCI America C30195ML

 View more versions of this product

Catalog No. C30195ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Cyclopentanecarboxaldehyde (stabilized with HQ)
Colorless
C6H10O
5 mL
cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde
C1CCC(C1)C=O
98.145
98.15
Liquid
872-53-7
40°C
MFCD00060798
1989
VELDYOPRLMJFIK-UHFFFAOYSA-N
cyclopentanecarbaldehyde
70106
≥97.0% (GC)

Chemical Identifiers

872-53-7
98.145
VELDYOPRLMJFIK-UHFFFAOYSA-N
70106
C1CCC(C1)C=O
C6H10O
MFCD00060798
cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde
cyclopentanecarbaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions