missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxaldehyde (stabilized with HQ) 90.0+%, TCI America™

Numéro de catalogue. C30191ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C30191ML 1 mL
C30195ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C30191ML Fournisseur TCI America Code fournisseur C30191ML
Il en reste null

Identifiants chimiques

CAS 872-53-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00060798
InChI Key VELDYOPRLMJFIK-UHFFFAOYSA-N
Synonym cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde
PubChem CID 70106
IUPAC Name cyclopentanecarbaldehyde
SMILES C1CCC(C1)C=O

Spécifications

Color Colorless
Boiling Point 40°C
Quantity 1 mL
UN Number 1989
Formula Weight 98.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentanecarboxaldehyde (stabilized with HQ)
TSCA No
Recommended Storage Refrigerator