missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentane 98.0+%, TCI America™

Catalog No. p-7124568
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C069925ML 25 mL
C0699100ML 100 mL
C0699500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C069925ML Supplier TCI America Supplier No. C069925ML
Only null left

Chemical Identifiers

CAS 287-92-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.135
MDL Number MFCD00001356
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
PubChem CID 9253
ChEBI CHEBI:23492
IUPAC Name cyclopentane
SMILES C1CCCC1

Specifications

Color Colorless
Boiling Point 50°C
Quantity 25 mL
UN Number 1146
Formula Weight 70.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopentane
EINECSNumber (3)-4166
RTECSNumber GY2390000
TSCA Yes