missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentane, 97%, extra pure

Catalog No. AC111480010
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111480010 1 L Glass bottle
AC111481000 100 mL Glass bottle
AC111480025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111480010 Supplier Thermo Scientific Chemicals Supplier No. 111480010

Cyclopentane, 97%, C5H10, CAS Number-287-92-3, ciclopentano, cyclopentan, cyclopentanes, cyclopentyl group, hsdb 62, hydrocarbons, cyclic c5 and c6, pentamethylene, t86pb90rnu, unii-t86pb90rnu, zyklopentan, 1L, 96% min. (GC), 05, 19, 50.0 deg.C, CHEBI:23492, White, 0.7500g/mL, 206-016-6, -7 deg.C | CAS: 287-92-3 | C5H10 | 70.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 287-92-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.15
MDL Number MFCD00001356
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
PubChem CID 9253
ChEBI CHEBI:23492
IUPAC Name cyclopentane
SMILES C1CCCC1

Specifications

Melting Point -94.0°C
Color White
Density 0.7500g/mL
Boiling Point 50.0°C
Flash Point −7°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4050 to 1.4070
Linear Formula C5H10
Quantity 1 L
Residue after Evaporation 0.01 % max.
Beilstein 05, 19
Specific Gravity 0.75
Merck Index 15, 2736
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,alcohol,ether and,most hydrocarbon solvents
Viscosity 0.44 mPa.s (20°C)
Formula Weight 70.15
Percent Purity 97%
Grade Extra Pure
Physical Form Powder
Chemical Name or Material Cyclopentane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
EINECSNumber 206-016-6

RUO – Research Use Only