Learn More
Cyclopentane, 95%
CAS: 287-92-3 | C5H10 | 70.135 g/mol
Supplier: Thermo Scientific Chemicals A146970F
Description
Cyclopentane is used as green blowing agent and involved in the production of polyurethane insulating foam, which is used in refrigerators, freezers, water heaters, construction panels, insulated pipes and roofs. As a lubricant, it finds applications in computer hard drives and outer space equipment due to its low volatile nature. It is widely useful in the preparation of resin, adhesives and pharmaceutical intermediate. It is an additive in gasoline. Since it is a halogen free compound and has zero-ozone depletion potential, it replaces the conventionally used chloro fluoro carbon (CFC) in refrigeration and thermal insulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclopentane is used as green blowing agent and involved in the production of polyurethane insulating foam, which is used in refrigerators, freezers, water heaters, construction panels, insulated pipes and roofs. As a lubricant, it finds applications in computer hard drives and outer space equipment due to its low volatile nature. It is widely useful in the preparation of resin, adhesives and pharmaceutical intermediate. It is an additive in gasoline. Since it is a halogen free compound and has zero-ozone depletion potential, it replaces the conventionally used chloro fluoro carbon (CFC) in refrigeration and thermal insulation.
Solubility
Miscible with ethanol, ether and acetone. Slightly miscible with water.
Notes
Avoid heat, direct sunlight, flames and sparks. Incompatible with strong oxidizing agents.
Specifications
| Cyclopentane | |
| -95°C | |
| 47°C to 49°C | |
| 95% | |
| C5H10 | |
| MFCD00001356 | |
| UN1146 | |
| 14,2741 | |
| Miscible with ethanol,ether and acetone. Slightly miscible with water. | |
| C1CCCC1 | |
| 70.135 | |
| CHEBI:23492 | |
| 95% |
| 287-92-3 | |
| 0.748 | |
| −37°C (−34°F) | |
| Petrol-like | |
| 1.406 | |
| 2500 mL | |
| 1900195 | |
| pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6 | |
| RGSFGYAAUTVSQA-UHFFFAOYSA-N | |
| cyclopentane | |
| 9253 | |
| 70.14 | |
| Liquid |
Chemical Identifiers
| 287-92-3 | |
| 70.135 | |
| RGSFGYAAUTVSQA-UHFFFAOYSA-N | |
| 9253 | |
| cyclopentane |
| C5H10 | |
| MFCD00001356 | |
| pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6 | |
| CHEBI:23492 | |
| C1CCCC1 |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
H225-H304-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CYCLOPENTANE
EINECSNumber : 206-016-6
RTECSNumber : GY2390000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only