Learn More
Cyclopentane, 95+%, pure
Cyclopentane, 95+%, C5H10, CAS Number-287-92-3, pentamethylene, cyclopentanes, hsdb 62, hydrocarbons, cyclic c5 and c6, cyclopentan, t86pb90rnu, unii-t86pb90rnu, ciclopentano, cyclopentyl group, zyklopentan, 5L, 95% min. (GC), 50.0 deg.C, CHEBI:23492, Colorless, 0.7500g/mL, 206-016-6, -7 deg.C | CAS: 287-92-3 | C5H10 | 70.15 g/mol
$318.39 - $1026.70
Chemical Identifiers
| CAS | 287-92-3 |
|---|---|
| Molecular Formula | C5H10 |
| Molecular Weight (g/mol) | 70.15 |
| MDL Number | MFCD00001356 |
| InChI Key | RGSFGYAAUTVSQA-UHFFFAOYSA-N |
| Synonym | pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6 |
| PubChem CID | 9253 |
| ChEBI | CHEBI:23492 |
| IUPAC Name | cyclopentane |
| SMILES | C1CCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC397430010
|
Thermo Scientific Chemicals
397430010 |
1 L |
Each for $318.39
|
|
|||||
|
AC397430050
|
Thermo Scientific Chemicals
397430050 |
5 L |
Each for $1,026.70
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 287-92-3 | |
| 70.15 | |
| RGSFGYAAUTVSQA-UHFFFAOYSA-N | |
| 9253 | |
| cyclopentane |
| C5H10 | |
| MFCD00001356 | |
| pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6 | |
| CHEBI:23492 | |
| C1CCCC1 |
Specifications
| 287-92-3 | |
| Colorless | |
| 50.0°C | |
| Authentic | |
| Glass Bottle | |
| 1.4050 to 1.4070 | |
| MFCD00001356 | |
| 0.001% max. | |
| 15, 2736 | |
| Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,alcohol,ether and,most hydrocarbon solvents | |
| C1CCCC1 | |
| 70.15 | |
| 0.44 mPa.s (20°C) | |
| 70.15 | |
| Pure | |
| Cyclopentane, Extra Dry |
| -94.0°C | |
| 0.7500g/mL | |
| −7°C | |
| 95% min. (GC) | |
| C5H10 | |
| C5H10 | |
| 1 L | |
| 0.75 | |
| pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6 | |
| RGSFGYAAUTVSQA-UHFFFAOYSA-N | |
| cyclopentane | |
| 9253 | |
| CHEBI:23492 | |
| 95+% | |
| Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
GHS Signal Word: Danger
EINECSNumber : 206-016-6
RUO – Research Use Only