Learn More
Cyclopentadienylmolybdenum tricarbonyl dimer, 98%
CAS: 12091-64-4 | C16H2Mo2O6 | 482.09 g/mol
$97.10 - $337.18
Chemical Identifiers
| CAS | 12091-64-4 |
|---|---|
| Molecular Formula | C16H2Mo2O6 |
| Molecular Weight (g/mol) | 482.09 |
| MDL Number | MFCD00009947 |
| InChI Key | AYBJYGWNVNZERR-UHFFFAOYSA-N |
| Synonym | Bis(tricarbonylcyclopentadienylmolybdenum); Tricarbonylcyclopentadienylmolybdenum dimer |
Description
It is widely used as a chemical intermediate in the laboratories, manufacture purposes, chemical and pharmaceutical intemediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12091-64-4 | |
| 482.09 | |
| AYBJYGWNVNZERR-UHFFFAOYSA-N |
| C16H2Mo2O6 | |
| MFCD00009947 | |
| Bis(tricarbonylcyclopentadienylmolybdenum); Tricarbonylcyclopentadienylmolybdenum dimer |
Specifications
| 12091-64-4 | |
| Purple to Red | |
| MFCD00009947 | |
| UN3466 | |
| Bis(tricarbonylcyclopentadienylmolybdenum); Tricarbonylcyclopentadienylmolybdenum dimer | |
| AYBJYGWNVNZERR-UHFFFAOYSA-N | |
| 490.13 | |
| Crystalline |
| 220°C (decomposition) | |
| C16H2Mo2O6 | |
| 1 g | |
| Air sensitive | |
| It is insoluble in water. | |
| 482.09 | |
| 98% | |
| Cyclopentadienylmolybdenum tricarbonyl dimer |
Safety and Handling
GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 235-156-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only