missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentadienylmolybdenum tricarbonyl dimer, 98%
Description
It is widely used as a chemical intermediate in the laboratories, manufacture purposes, chemical and pharmaceutical intemediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 220°C (decomposition) |
| Color | Purple to Red |
| Quantity | 1 g |
| UN Number | UN3466 |
| Sensitivity | Air sensitive |
| Solubility Information | It is insoluble in water. |
| Formula Weight | 490.13 |
| Percent Purity | 98% |
| Physical Form | Crystalline |
| Chemical Name or Material | Cyclopentadienylmolybdenum tricarbonyl dimer |
Safety and Handling
| Hazard Category | H302+H312+H332 |
| Hazard Statement | GHS H Statement H300-H330 Fatal if swallowed. Fatal if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 235-156-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only