missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentadienyl Thallium 95.0+%, TCI America™
$285.21 - $678.52
Chemical Identifiers
| CAS | 34822-90-7 |
|---|---|
| Molecular Formula | C5H5Tl |
| MDL Number | MFCD00134141 |
Chemical Identifiers
| 34822-90-7 | |
| C5H5Tl | |
| MFCD00134141 |
Specifications
| 34822-90-7 | |
| 1 g | |
| MFCD00134141 | |
| 269.48 | |
| Cyclopentadienyl Thallium |
| Crystalline Powder | |
| C5H5Tl | |
| 1707 | |
| ≥95.0% (T) |
Safety and Handling
TSCA : Yes