missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Cycloolefin Microplate Lid
GREENER_CHOICE

Catalog No. 07001053
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
07001053 Sterile
07000933 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07001053 Supplier Greiner Bio-One Supplier No. 792891ST

For use with 1536-Well Cycloolefin Low-Evaporation Clear Microplate.

  • Universal fit
  • Clear
  • Sterile

Specifications

Color Clear
Material Cycloolefin
Sterility Sterile
Product Type Universal Lid
Plate Compatibility 1536-well Cycloolefin Low-evaporation Clear Microplate