missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclooctylamine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -48°C |
| Color | Yellow |
| Boiling Point | 190°C |
| Quantity | 25 mL |
| UN Number | 2735 |
| Formula Weight | 127.23 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Cyclooctylamine |
Safety and Handling
| TSCA | Yes |