missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctene 95.0+%, TCI America™

Numéro de catalogue. C0506500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C0506500ML 500 mL
C050625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C0506500ML Fournisseur TCI America Code fournisseur C0506500ML
Only null left

Chemical Identifiers

CAS 931-88-4
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00001753
InChI Key URYYVOIYTNXXBN-UPHRSURJSA-N
PubChem CID 638079
IUPAC Name cyclooctene
SMILES C1CCCC=CCC1

Specifications

Color Colorless
Boiling Point 144°C
Quantity 500 mL
UN Number 3295
Formula Weight 110.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclooctene