missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclooctene 95.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 144°C |
| Quantity | 25 mL |
| UN Number | 3295 |
| Formula Weight | 110.20 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Cyclooctene |
Safety and Handling
| TSCA | Yes |