missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclooctanone 97.0+%, TCI America™
$29.83 - $52.19
Chemical Identifiers
| CAS | 502-49-8 |
|---|---|
| Molecular Formula | C8H14O |
| Molecular Weight (g/mol) | 126.199 |
| MDL Number | MFCD00001754 |
| InChI Key | IIRFCWANHMSDCG-UHFFFAOYSA-N |
| Synonym | cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference |
| PubChem CID | 10403 |
| IUPAC Name | cyclooctanone |
| SMILES | C1CCCC(=O)CCC1 |
Chemical Identifiers
| 502-49-8 | |
| 126.199 | |
| IIRFCWANHMSDCG-UHFFFAOYSA-N | |
| 10403 | |
| C1CCCC(=O)CCC1 |
| C8H14O | |
| MFCD00001754 | |
| cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference | |
| cyclooctanone |
Specifications
| 502-49-8 | |
| White | |
| MFCD00001754 | |
| cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference | |
| C1CCCC(=O)CCC1 | |
| 126.199 | |
| 126.20 | |
| Crystalline Lumps |
| 45°C | |
| C8H14O | |
| 10 g | |
| IIRFCWANHMSDCG-UHFFFAOYSA-N | |
| cyclooctanone | |
| 10403 | |
| ≥97.0% (GC) | |
| Cyclooctanone |
Safety and Handling
RTECSNumber : GX9800000
TSCA : Yes