missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanone 97.0+%, TCI America™

Catalog No. C050410G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C050410G 10 g
C050425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C050410G Supplier TCI America Supplier No. C050410G
Only null left

Chemical Identifiers

CAS 502-49-8
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00001754
InChI Key IIRFCWANHMSDCG-UHFFFAOYSA-N
Synonym cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
PubChem CID 10403
IUPAC Name cyclooctanone
SMILES C1CCCC(=O)CCC1

Specifications

Melting Point 45°C
Color White
Quantity 10 g
Formula Weight 126.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Cyclooctanone
RTECSNumber GX9800000
TSCA Yes