missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanone, 98%

CAS: 502-49-8 | C8H14O | 126.199 g/mol

Supplier:  Thermo Scientific Chemicals B2091122

 View more versions of this product

Catalog No. AAB2091122


Only null left
Explore available promotions

Description

Description

Cyclooctanone is a aliphatic cycle that exhibited inhibitory activity towards aldosterone synthase, a promising therapeutic target for the treatment of cardiovascular diseases related to abnormally high aldosterone level. It is also used in the synthesis of 14-membered lactones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water at 20°C 15g/L. Soluble in acetone, alcohol, chloroform, methanol and benzene.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
Specifications

Specifications

Cyclooctanone
37°C to 44°C
195°C to 197°C
Pleasant
MFCD00001754
UN1759
cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
IIRFCWANHMSDCG-UHFFFAOYSA-N
cyclooctanone
10403
98%
502-49-8
0.96
72°C (161°F)
C8H14O
100 g
1280738
Soluble in water at 20°C 15g/L. Soluble in acetone,alcohol,chloroform,methanol and benzene.
C1CCCC(=O)CCC1
126.199
126.2

Chemical Identifiers

502-49-8
126.199
IIRFCWANHMSDCG-UHFFFAOYSA-N
10403
C1CCCC(=O)CCC1
C8H14O
MFCD00001754
cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
cyclooctanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

EINECSNumber : 207-940-2

RTECSNumber : GX9800000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only