missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanone, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 111441000

Catalog No. AC111441000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Cyclooctanone
502-49-8
195.0°C to 197.0°C
Authentic
Glass bottle
C8H14(=O)
100g
0.95
Solubility in water: 15g/l (20°C). Other solubilities: soluble in alcohol,acetone and benzene
C1CCCC(=O)CCC1
126.2
126.2
98%
0.9500g/mL
74°C
98%
C8H14O
MFCD00001754
07, 21
cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
IIRFCWANHMSDCG-UHFFFAOYSA-N
cyclooctanone
10403
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-940-2

RTECSNumber : GX9800000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only