missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanone, 98%

CAS: 502-49-8 | C8H14O | 126.2 g/mol

$75.09 - $75.09

Chemical Identifiers

CAS 502-49-8
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.2
MDL Number MFCD00001754
InChI Key IIRFCWANHMSDCG-UHFFFAOYSA-N
Synonym cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
PubChem CID 10403
IUPAC Name cyclooctanone
SMILES C1CCCC(=O)CCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC111440250
SDS View Documents
Thermo Scientific Chemicals
111440250
25 g
Each for $75.09
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

502-49-8
126.2
IIRFCWANHMSDCG-UHFFFAOYSA-N
10403
C1CCCC(=O)CCC1
C8H14O
MFCD00001754
cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
cyclooctanone

Specifications

502-49-8
Colorless to White
195.0°C to 197.0°C
Authentic
Glass bottle
C8H14(=O)
25 g
0.95
Solubility in water: 15g/l (20°C). Other solubilities: soluble in alcohol,acetone and benzene
C1CCCC(=O)CCC1
126.2
126.2
Crystalline Low Melting Solid
39.0°C to 44.0°C
0.9500g/mL
74°C
98%
C8H14O
MFCD00001754
07, 21
cyclooctan-1-one, cyclooctyloxy, chembl18737, cyclooctanon, oxocyclooctane, cyclo-octanone, cyclooctanone, acmc-209kku, wln: l8vtj, 4-07-00-00049 beilstein handbook reference
IIRFCWANHMSDCG-UHFFFAOYSA-N
cyclooctanone
10403
98%
Cyclooctanone, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-940-2

RTECSNumber : GX9800000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only