missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanol 98.0+%, TCI America™

Catalog No. C062325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C062325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C062325G Supplier TCI America Supplier No. C062325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cyclooctanol is a cyclic secondary alcohol suitable for use as a chemical intermediate in organic synthesis.

Chemical Identifiers

CAS 696-71-9
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00001744
InChI Key FHADSMKORVFYOS-UHFFFAOYSA-N
Synonym cyclooctyl alcohol, 1-hydroxycyclooctane, acmc-209oac, fhadsmkorvfyos-uhfffaoysa
PubChem CID 12766
IUPAC Name cyclooctanol
SMILES OC1CCCCCCC1

Specifications

Color Colorless
Boiling Point 108°C
Quantity 25 g
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclooctanol