missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanol, 97%

Catalog No. AAA1782514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1782514 25 g
AAA1782522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1782514 Supplier Thermo Scientific Chemicals Supplier No. A1782514

CAS: 696-71-9 | C8H16O | 128.22 g/mol

Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.

Solubility
Miscible with acetone.

Notes
Contains 75+% cyclopentane; remainder consists of C{5} and C{6} hydrocarbon.Incompatible with strong oxidizing agents, acid chloride, and acid anhydrides.

Chemical Identifiers

CAS 696-71-9
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00001744
InChI Key FHADSMKORVFYOS-UHFFFAOYSA-N
Synonym cyclooctyl alcohol, 1-hydroxycyclooctane, acmc-209oac, fhadsmkorvfyos-uhfffaoysa
PubChem CID 12766
IUPAC Name cyclooctanol
SMILES OC1CCCCCCC1

Specifications

Melting Point 14°C to 15°C
Density 0.97
Boiling Point 97°C to 98°C (11 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.486
Quantity 25 g
Beilstein 1848166
Solubility Information Miscible with acetone.
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material Cyclooctanol
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H301-H227-H315-H319
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 211-800-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only