missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctane, 99+%

CAS: 292-64-8 | C8H16 | 112.216 g/mol

$74.51 - $190.66

Chemical Identifiers

CAS 292-64-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00004162
InChI Key WJTCGQSWYFHTAC-UHFFFAOYSA-N
Synonym octamethylene, cyclooctan, unii-kkz3kbs654, kkz3kbs654, cyclooctane, acmc-209h7x, cambridge id 6337558, cambridge id 6511221, cambridge id 6544558, cyclooctane, analytical standard
PubChem CID 9266
IUPAC Name cyclooctane
SMILES C1CCCCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL15000AD
View Documents
Thermo Scientific Chemicals
L15000AD
50 mL N/A
Only null left
 
AAL15000AK
View Documents
Thermo Scientific Chemicals
L15000AK
250 mL N/A
Only null left
 
Description

Description

Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

292-64-8
112.216
WJTCGQSWYFHTAC-UHFFFAOYSA-N
9266
C1CCCCCCC1
C8H16
MFCD00004162
octamethylene, cyclooctan, unii-kkz3kbs654, kkz3kbs654, cyclooctane, acmc-209h7x, cambridge id 6337558, cambridge id 6511221, cambridge id 6544558, cyclooctane, analytical standard
cyclooctane

Specifications

292-64-8
0.835
28°C (82°F)
1.458
50 mL
1900349
Not miscible or difficult to mix in water.
C1CCCCCCC1
112.216
112.22
Cyclooctane
12°C to 14°C
150°C to 152°C
C8H16
MFCD00004162
UN3295
octamethylene, cyclooctan, unii-kkz3kbs654, kkz3kbs654, cyclooctane, acmc-209h7x, cambridge id 6337558, cambridge id 6511221, cambridge id 6544558, cyclooctane, analytical standard
WJTCGQSWYFHTAC-UHFFFAOYSA-N
cyclooctane
9266
≥99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H226-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 206-031-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only