missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyldiphenylphosphine 97.0+%, TCI America™

Catalog No. p-7124974
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14295G 5 g
C142925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14295G Supplier TCI America Supplier No. C14295G
Only null left

Chemical Identifiers

CAS 6372-42-5
Molecular Formula C18H21P
Molecular Weight (g/mol) 268.34
MDL Number MFCD00046360
InChI Key ZXKWUYWWVSKKQZ-UHFFFAOYSA-N
Synonym cyclohexyldiphenylphosphine, phosphine, cyclohexyldiphenyl, cyclohexyl diphenyl phosphine, diphenylcyclohexylphosphine, diphenylphosphinocyclohexane, cyclohexyl-diphenyl-phosphane, pubchem6451, acmc-1ba4g, cyclohexyl diphenyl phosphane
PubChem CID 80756
IUPAC Name cyclohexyldiphenylphosphane
SMILES C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 60°C
Color White
Quantity 5 g
Formula Weight 268.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexyldiphenylphosphine