missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyldiphenylphosphine, 98%

CAS: 6372-42-5 | C18H21P | 268.34 g/mol

$284.50 - $284.50

Chemical Identifiers

CAS 6372-42-5
Molecular Formula C18H21P
Molecular Weight (g/mol) 268.34
MDL Number MFCD00046360
InChI Key ZXKWUYWWVSKKQZ-UHFFFAOYSA-N
Synonym cyclohexyldiphenylphosphine, phosphine, cyclohexyldiphenyl, cyclohexyl diphenyl phosphine, diphenylcyclohexylphosphine, diphenylphosphinocyclohexane, cyclohexyl-diphenyl-phosphane, pubchem6451, acmc-1ba4g, cyclohexyl diphenyl phosphane
PubChem CID 80756
IUPAC Name cyclohexyl(diphenyl)phosphane
SMILES C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC406100050
SDS View Documents
Thermo Scientific Chemicals
406100050
5 g Glass bottle
Each for $284.50
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6372-42-5
268.34
ZXKWUYWWVSKKQZ-UHFFFAOYSA-N
80756
C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1
C18H21P
MFCD00046360
cyclohexyldiphenylphosphine, phosphine, cyclohexyldiphenyl, cyclohexyl diphenyl phosphine, diphenylcyclohexylphosphine, diphenylphosphinocyclohexane, cyclohexyl-diphenyl-phosphane, pubchem6451, acmc-1ba4g, cyclohexyl diphenyl phosphane
cyclohexyl(diphenyl)phosphane

Specifications

6372-42-5
White
Authentic
Glass bottle
MFCD00046360
cyclohexyldiphenylphosphine, phosphine, cyclohexyldiphenyl, cyclohexyl diphenyl phosphine, diphenylcyclohexylphosphine, diphenylphosphinocyclohexane, cyclohexyl-diphenyl-phosphane, pubchem6451, acmc-1ba4g, cyclohexyl diphenyl phosphane
ZXKWUYWWVSKKQZ-UHFFFAOYSA-N
cyclohexyl(diphenyl)phosphane
80756
98%
Cyclohexyldiphenylphosphine
53.0°C
>110°C
98%
C18H21P
5 g
Solubility in water: insoluble in water
C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1
268.34
268.33
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-904-2

RTECSNumber : SY8387500

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only