missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine 99.0+%, TCI America™

Catalog No. C049425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C049425ML 25 mL
C0494500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C049425ML Supplier TCI America Supplier No. C049425ML

Chemical Identifiers

CAS 108-91-8
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00001486
InChI Key PAFZNILMFXTMIY-UHFFFAOYSA-N
Synonym cyclohexylamine, aminocyclohexane, hexahydroaniline, hexahydrobenzenamine, aminohexahydrobenzene, cyclohexyl amine, 1-cyclohexylamine, 1-aminocyclohexane, aniline, hexahydro, benzenamine, hexahydro
PubChem CID 7965
ChEBI CHEBI:15773
IUPAC Name cyclohexanamine
SMILES C1CCC(CC1)N

Specifications

Melting Point 18°C
Color Colorless
Boiling Point 135°C
Quantity 25 mL
UN Number 2357
Formula Weight 99.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexylamine
EINECSNumber (3)-2258
RTECSNumber GX0700000
TSCA Yes