missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexylamine Hydrochloride 98.0+%, TCI America™
$34.66 - $190.59
Chemical Identifiers
| CAS | 4998-76-9 |
|---|---|
| Molecular Formula | C6H14ClN |
| Molecular Weight (g/mol) | 135.64 |
| MDL Number | MFCD00050540 |
| InChI Key | ZJUGSKJHHWASAF-UHFFFAOYSA-N |
| Synonym | cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361 |
| PubChem CID | 2723770 |
| IUPAC Name | cyclohexanaminium chloride |
| SMILES | [Cl-].[NH3+]C1CCCCC1 |
Chemical Identifiers
| 4998-76-9 | |
| 135.64 | |
| ZJUGSKJHHWASAF-UHFFFAOYSA-N | |
| 2723770 | |
| [Cl-].[NH3+]C1CCCCC1 |
| C6H14ClN | |
| MFCD00050540 | |
| cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361 | |
| cyclohexanaminium chloride |
Specifications
| 4998-76-9 | |
| C6H14ClN | |
| 25 g | |
| ZJUGSKJHHWASAF-UHFFFAOYSA-N | |
| cyclohexanaminium chloride | |
| 2723770 | |
| ≥98.0% (N) | |
| Cyclohexylamine Hydrochloride |
| White | |
| MFCD00050540 | |
| cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361 | |
| [Cl-].[NH3+]C1CCCCC1 | |
| 135.64 | |
| 135.64 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : GX1440000
TSCA : Yes