missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine Hydrochloride 98.0+%, TCI America™

Catalog No. C0495500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0495500G 500 g
C049525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0495500G Supplier TCI America Supplier No. C0495500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4998-76-9
Molecular Formula C6H14ClN
Molecular Weight (g/mol) 135.64
MDL Number MFCD00050540
InChI Key ZJUGSKJHHWASAF-UHFFFAOYSA-N
Synonym cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361
PubChem CID 2723770
IUPAC Name cyclohexanaminium chloride
SMILES [Cl-].[NH3+]C1CCCCC1

Specifications

Color White
Quantity 500 g
Formula Weight 135.64
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Cyclohexylamine Hydrochloride
RTECSNumber GX1440000
TSCA Yes