missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexylamine Hydrochloride, 99%, Spectrum™ Chemical
Specifications
Specifications
| Physical Form | Flakes or Powder |
| CAS Max % | 1 |
| Quantity | 500 g |
| Packaging | Amber Glass Bottle |
| Molecular Weight (g/mol) | 135.63 |
| Percent Purity | 99% |
| Grade | Ungraded |
| Chemical Name or Material | Cyclohexylamine Hydrochloride |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.