missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine Hydrochloride, 99%, Spectrum™ Chemical

Catalog No. 18610120
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610120 25 g Amber Glass Bottle
18610117 100 g Amber Glass Bottle
18610121 500 g Amber Glass Bottle
18610119 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18610120 Supplier Spectrum Chemical Mfg Cor Supplier No. C212725GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 4998-76-9

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.