missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine Hydrochloride, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor C212725KGBL

 View more versions of this product

Catalog No. 18610119

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4998-76-9
25 kg
Reagent
1
99%
Fiber Drum

Chemical Identifiers

4998-76-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.