missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine hydrochloride, 98+%

CAS: 4998-76-9 | C6H14ClN | 135.64 g/mol

$69.73 - $229.49

Chemical Identifiers

CAS 4998-76-9
Molecular Formula C6H14ClN
Molecular Weight (g/mol) 135.64
MDL Number MFCD00050540
InChI Key ZJUGSKJHHWASAF-UHFFFAOYSA-N
Synonym cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361
PubChem CID 2723770
IUPAC Name cyclohexanamine;hydrochloride
SMILES [Cl-].[NH3+]C1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1730822
View Documents
Thermo Scientific Chemicals
A1730822
100 g N/A
Only null left
 
AAA1730836
View Documents
Thermo Scientific Chemicals
A1730836
500 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4998-76-9
135.64
ZJUGSKJHHWASAF-UHFFFAOYSA-N
2723770
[Cl-].[NH3+]C1CCCCC1
C6H14ClN
MFCD00050540
cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361
cyclohexanamine;hydrochloride

Specifications

4998-76-9
Odorless
MFCD00050540
3613002
cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361
[Cl-].[NH3+]C1CCCCC1
135.64
135.64
Cyclohexylamine hydrochloride
206°C to 209°C
C6H14ClN
100 g
Hygroscopic
ZJUGSKJHHWASAF-UHFFFAOYSA-N
cyclohexanamine;hydrochloride
2723770
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H361-H315-H319-H335
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H318-H335

EINECSNumber : 225-661-4

RTECSNumber : GX1440000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only