missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine hydrochloride, 98+%

Catalog No. AAA1730836
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1730836 500 g
AAA1730822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1730836 Supplier Thermo Scientific Chemicals Supplier No. A1730836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4998-76-9 | C6H14ClN | 135.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4998-76-9
Molecular Formula C6H14ClN
Molecular Weight (g/mol) 135.64
MDL Number MFCD00050540
InChI Key ZJUGSKJHHWASAF-UHFFFAOYSA-N
Synonym cyclohexylamine hydrochloride, cyclohexanamine hydrochloride, cyclohexanamine, hydrochloride, aminocyclohexane hydrochloride, cyclohexylamine.hcl, unii-tn6vyf8thc, dsstox_cid_361, cyclohexanamine, hydrochloride 1:1, dsstox_rid_75539, dsstox_gsid_20361
PubChem CID 2723770
IUPAC Name cyclohexanamine;hydrochloride
SMILES [Cl-].[NH3+]C1CCCCC1

Specifications

Melting Point 206°C to 209°C
Odor Odorless
Quantity 500 g
Beilstein 3613002
Sensitivity Hygroscopic
Formula Weight 135.64
Percent Purity ≥98%
Chemical Name or Material Cyclohexylamine hydrochloride
Hazard Category H302+H312+H332-H315-H318-H335
Hazard Statement GHS H Statement
H301-H361-H315-H319-H335
Toxic if swallowed.
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-661-4
RTECSNumber GX1440000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only