Learn More
Cyclohexylamine, 98+%
CAS: 108-91-8 | C6H13N | 99.177 g/mol
Supplier: Thermo Scientific Chemicals A158510F
Description
Cyclohexylamine is a useful as an intermediate in organic synthesis. It acts as a precursor to prepare pendant-armed dialdehydes, bromohexine HCl and sulfenamide-based reagents. It is widely used in the production of vulcanization accelerators like N-cyclohexyl-2-benzothiazole sulfonamide (CBS) and artificial sweeteners (cyclamates). As a corrosion inhibitor, it is used in the treatment of boiler water and prevents corrosion of metal surfaces that arise due to dissolved carbon dioxide. It is involved as a solvent in the synthesis of an inorganic-organic hybrid semiconductor, ZnS/CHA (CHA = cyclohexylamine) nanocomposites.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclohexylamine is a useful as an intermediate in organic synthesis. It acts as a precursor to prepare pendant-armed dialdehydes, bromohexine HCl and sulfenamide-based reagents. It is widely used in the production of vulcanization accelerators like N-cyclohexyl-2-benzothiazole sulfonamide (CBS) and artificial sweeteners (cyclamates). As a corrosion inhibitor, it is used in the treatment of boiler water and prevents corrosion of metal surfaces that arise due to dissolved carbon dioxide. It is involved as a solvent in the synthesis of an inorganic-organic hybrid semiconductor, ZnS/CHA (CHA = cyclohexylamine) nanocomposites.
Solubility
Miscible with water, ethanol, ethers, acetone, alcohol and ketones.
Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, carbon dioxide, sodium hypochlorite, organic acids, mineral acids and peroxides.
Specifications
| Cyclohexylamine | |
| -18°C | |
| 133°C to 134°C | |
| C6H13N | |
| MFCD00001486 | |
| UN2357 | |
| Air Sensitive | |
| cyclohexylamine, aminocyclohexane, hexahydroaniline, hexahydrobenzenamine, aminohexahydrobenzene, cyclohexyl amine, 1-cyclohexylamine, 1-aminocyclohexane, aniline, hexahydro, benzenamine, hexahydro | |
| PAFZNILMFXTMIY-UHFFFAOYSA-N | |
| cyclohexanamine | |
| 7965 | |
| 99.18 |
| 108-91-8 | |
| 0.868 | |
| 27°C (81°F) | |
| 1.4585 | |
| 2500 mL | |
| 471175 | |
| 14,2729 | |
| Miscible with water,ethanol,ethers,acetone,alcohol and ketones. | |
| C1CCC(CC1)N | |
| 99.177 | |
| CHEBI:15773 | |
| ≥98% |
Chemical Identifiers
| 108-91-8 | |
| 99.177 | |
| PAFZNILMFXTMIY-UHFFFAOYSA-N | |
| 7965 | |
| cyclohexanamine |
| C6H13N | |
| MFCD00001486 | |
| cyclohexylamine, aminocyclohexane, hexahydroaniline, hexahydrobenzenamine, aminohexahydrobenzene, cyclohexyl amine, 1-cyclohexylamine, 1-aminocyclohexane, aniline, hexahydro, benzenamine, hexahydro | |
| CHEBI:15773 | |
| C1CCC(CC1)N |
Safety and Handling
GHS H Statement
H314-H226-H361-H302-H312
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H301+H311-H314-H361f
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CYCLOHEXYLAMINE
EINECSNumber : 203-629-0
RTECSNumber : GX0700000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only