missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Cyclohexylamine
Description
Cyclohexylamine
Specifications
Specifications
| CAS | 108-91-8 |
| Melting Point | -17°C (lit.) |
| Density | 0.867 g/mL (at 25°C (literature)) |
| Boiling Point | 134°C (lit.) |
| Molecular Formula | C6H13N |
| Refractive Index | n20/D 1.459 (literature) |
| Linear Formula | C6H11NH2 |
| MDL Number | MFCD00001486 |
| Quantity | 100 mL |
| Synonym | Aminocyclohexane |
| Show More |