missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexyl Vinyl Ether (stabilized with KOH) 98.0+%, TCI America™
$108.11 - $710.21
Chemical Identifiers
| CAS | 2182-55-0 |
|---|---|
| Molecular Formula | C8H14O |
| Molecular Weight (g/mol) | 126.199 |
| MDL Number | MFCD00046358 |
| InChI Key | VGIYPVFBQRUBDD-UHFFFAOYSA-N |
| Synonym | cyclohexyl vinyl ether, vinyloxy cyclohexane, cyclohexane, ethenyloxy, vinylcyclohexyl ether, cyclohexylvinylether, vinyl cyclohexyl ether, ethenyloxy cyclohexane, vinyloxycyclohexane, cyclohexylvinyl ether, vinyloxy cyclohexane # |
| PubChem CID | 75129 |
| IUPAC Name | ethenoxycyclohexane |
| SMILES | C=COC1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C088525ML
|
TCI America
C088525ML |
25 mL |
Each for $108.11
|
|
|||||
|
C0885500ML
|
TCI America
C0885500ML |
500 mL |
Each for $710.21
|
|
|||||
Chemical Identifiers
| 2182-55-0 | |
| 126.199 | |
| VGIYPVFBQRUBDD-UHFFFAOYSA-N | |
| 75129 | |
| C=COC1CCCCC1 |
| C8H14O | |
| MFCD00046358 | |
| cyclohexyl vinyl ether, vinyloxy cyclohexane, cyclohexane, ethenyloxy, vinylcyclohexyl ether, cyclohexylvinylether, vinyl cyclohexyl ether, ethenyloxy cyclohexane, vinyloxycyclohexane, cyclohexylvinyl ether, vinyloxy cyclohexane # | |
| ethenoxycyclohexane |
Specifications
| 2182-55-0 | |
| 152°C | |
| MFCD00046358 | |
| 3271 | |
| VGIYPVFBQRUBDD-UHFFFAOYSA-N | |
| ethenoxycyclohexane | |
| 75129 | |
| ≥98.0% (GC) | |
| Cyclohexyl Vinyl Ether (stabilized with KOH) |
| Yellow | |
| C8H14O | |
| 25 mL | |
| cyclohexyl vinyl ether, vinyloxy cyclohexane, cyclohexane, ethenyloxy, vinylcyclohexyl ether, cyclohexylvinylether, vinyl cyclohexyl ether, ethenyloxy cyclohexane, vinyloxycyclohexane, cyclohexylvinyl ether, vinyloxy cyclohexane # | |
| C=COC1CCCCC1 | |
| 126.199 | |
| 126.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-3687
TSCA : Yes