missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. M0594500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0594500ML 500 mL
M059425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0594500ML Supplier TCI America Supplier No. M0594500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101-43-9
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00014292
InChI Key OIWOHHBRDFKZNC-UHFFFAOYSA-N
Synonym cyclohexyl methacrylate, methacrylic acid, cyclohexyl ester, 2-propenoic acid, 2-methyl-, cyclohexyl ester, unii-5l9uuv9t6q, 2-methyl-2-propenoic acid cyclohexyl ester, 5l9uuv9t6q, methacrylic acid cyclohexyl ester, ageflex chma, c-hma, cyclo hexyl methacrylate
PubChem CID 7561
IUPAC Name cyclohexyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1CCCCC1

Specifications

Color Yellow
Boiling Point 72°C
Quantity 500 mL
Formula Weight 168.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyl Methacrylate (stabilized with MEHQ)
EINECSNumber (3)-2305
TSCA Yes