missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl methacrylate, 97%, stab. with ca 50ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog No. AAL1063414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1063414 25 g
AAL1063422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1063414 Supplier Thermo Scientific Chemicals Supplier No. L1063414

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101-43-9
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00014292
InChI Key OIWOHHBRDFKZNC-UHFFFAOYSA-N
Synonym cyclohexyl methacrylate, methacrylic acid, cyclohexyl ester, 2-propenoic acid, 2-methyl-, cyclohexyl ester, unii-5l9uuv9t6q, 2-methyl-2-propenoic acid cyclohexyl ester, 5l9uuv9t6q, methacrylic acid cyclohexyl ester, ageflex chma, c-hma, cyclo hexyl methacrylate
PubChem CID 7561
IUPAC Name cyclohexyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1CCCCC1

Specifications

Density 0.97
Boiling Point 72°C to 73°C (5 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.4577
Quantity 25 g
Beilstein 2045235
Formula Weight 168.24
Percent Purity 97%
Chemical Name or Material Cyclohexyl methacrylate, stabilized with ≈50ppm 4-methoxyphenol
Hazard Category H227
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 202-943-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only