missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl(dimethoxy)methylsilane 98.0+%, TCI America™

Catalog No. C198225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C198225ML 25 mL
C1982500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C198225ML Supplier TCI America Supplier No. C198225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17865-32-6
Molecular Formula C9H20O2Si
Molecular Weight (g/mol) 188.34
MDL Number MFCD00071752
InChI Key XRLIOAUFFCHSIC-UHFFFAOYSA-N
Synonym cyclohexyldimethoxy methyl silane, cyclohexyldimethoxymethylsilane, cyclohexyl dimethoxy methylsilane, cyclohexylmethyldimethoxysilane, methylcyclohexyldimethoxysilane, cyclohexane, dimethoxymethylsilyl, cyclohexyl methyl dimethoxy silane, cyclohexyldimethoxymethyl silane, chms 112, cyclohexyl methyl dimethoxysilane
PubChem CID 569301
IUPAC Name (cyclohexylmethyl)dimethoxysilane
SMILES CO[SiH](CC1CCCCC1)OC

Specifications

Color Yellow
Boiling Point 196°C
Quantity 25 mL
UN Number 3082
Formula Weight 188.34
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclohexyl(dimethoxy)methylsilane
EINECSNumber (3)-4027
TSCA Yes